Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 405)

Supplier: AAT BIOQUEST
10052020 10052021
AATB10052020EA 328 GBP
AATB10052020 AATB10052021
Anti-CD5 Mouse Monoclonal Antibody [clone: UCHT2] (iFluor® 405)
Antibodies
UCHT2 is an anti-human monoclonal antibody that is specific for the CD5 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Caution: For research use only.

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: UCHT2
Conjugation: iFluor® 405
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR