Methyl yellow, Supelco®

Supplier: Merck
Danger

Synonyms: Dimethyl yellow, Sudan Yellow, 4-(Dimethylamino)azobenzene, 4-(N,N-Dimethylamino)azobenzene

M-Clarity™ quality level = MQ100

73225-25MG
73225-25MGEA 136 GBP
73225-25MG
Methyl yellow, Supelco®
Methyl Yellow
Formula: C₁₄H₁₅N₃
MW: 225,29 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 113…117 °C
Density: 1,2233 g/cm³ (20 °C)
MDL Number: MFCD00008308
CAS Number: 60-11-7
EINECS: 200-455-7
UN: 2811
ADR: 6.1,III
Merck Index: 12,03279
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=167c456c2cf6a92e07e9e5b0a43e0d74604beb99&vid=167c456c2cf6a92e07e9e5b0a43e0d74604beb99&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR