1-(4-Pyridyl)piperazine

Supplier: Molekula

Synonyms: 1-(Pyridin-4-yl)piperazine4-(1-Piperazinyl)pyridine

MOLEM81320264EA 0 GBP
MOLEM81320264
1-(4-Pyridyl)piperazine
1-(4-Pyridyl)piperazine
Formula: C₉H₁₃N₃
MW: 163,22 g/mol
Boiling Pt: 195…200 °C (0,3 mmHg)
Melting Pt: 140 °C
MDL Number: MFCD00040745
CAS Number: 1008-91-9
EINECS: 213-764-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR