Methyl yellow

Supplier: Molekula
Danger

Synonyms: Dimethyl yellowSudan Yellow4-(Dimethylamino)azobenzene4-(N,N-Dimethylamino)azobenzene

MOLE27793472-25GEA 68.9 GBP
MOLE27793472-25G
Methyl yellow
Methyl Yellow
Formula: C₁₄H₁₅N₃
MW: 225,29 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 113…117 °C
Density: 1,2233 g/cm³ (20 °C)
MDL Number: MFCD00008308
CAS Number: 60-11-7
EINECS: 200-455-7
UN: 2811
ADR: 6.1,III
Merck Index: 12,03279

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR