5-Amino-1H-tetrazole monohydrate

Supplier: Molekula

Synonyms: 1H-Tetrazol-5-ylamine monohydrate5-Aminotetrazole monohydrate1H-Tetrazol-5-amine hydrate

MOLE35827584-100GEA 138 GBP
MOLE35827584-100G
5-Amino-1H-tetrazole monohydrate
5-Amino-1H-tetrazole monohydrate
Formula: CH₃N₅·1H₂O
MW: 103,08 g/mol
Melting Pt: 216…217 °C
Density: 1,51 g/cm³ (20 °C)
MDL Number: MFCD00149327
CAS Number: 15454-54-3
EINECS: 224-581-7
UN: 1325
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR