3-Chloropropyl isocyanate ≥97%
Supplier: Thermo Scientific
|
Danger
|
L15993.03
L15993.06
L15993.03EA
23.1
GBP
L15993.03
L15993.06
3-Chloropropyl isocyanate ≥97%
3-Chloropropyl isocyanate
Purity: 97%
CAS No.: 13010-19-0
Molecular Formula: Cl(CH2)3NCO
Formula Weight: 119.55
Storage Temperature: 2°C to 8°C
Flash Point: 61°(141°F)
Refractive Index: 1.4520
Odour: Pungent
Boiling Point: 152-154°
Beilstein Reference: 1098789
UN No.: 2206
MDL No.: MFCD00010384
Formula:
C₄H₆ClNO MW: 119,55 g/mol Boiling Pt: 152…154 °C Density: 1,165 Flash Pt: 61 °C (141 °F) |
MDL Number:
MFCD00010384 CAS Number: 13010-19-0 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...