3-Thiopheneethanol ≥98%
Supplier: Thermo Scientific
Warning
|
Synonyms:
Thiophen-3-ethanol, 2-(Thiophen-3-yl)ethanol, 2-(3-Thienyl)ethanol
L15138.03
L15138.06
L15138.03EA
22
GBP
L15138.03
L15138.06
3-Thiopheneethanol ≥98%
3-Thiopheneethanol
Purity: 98%
CAS No.: 13781-67-4
Molecular Formula: C6H8OS
Formula Weight: 128.19
Flash Point: 92°(197°F)
Refractive Index: 1.5530
Boiling Point: 110-111°/14mm
Beilstein Reference: 107080
MDL No.: MFCD00009766
Formula:
C₆H₈OS MW: 128,19 g/mol Boiling Pt: 110…111 °C (14 mmHg) Density: 1,159 Flash Pt: 92 °C (197 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009766 CAS Number: 13781-67-4 EINECS: 237-434-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...