N-Butylethylenediamine 97%
Supplier: Thermo Fisher Scientific
|
Danger
|
L13437.06
L13437.14
L13437.06EA
78.6
GBP
L13437.06
L13437.14
N-Butylethylenediamine 97%
N-Butylethylenediamine
Purity: 97%
CAS No.: 19522-69-1
Molecular Formula: CH3(CH2)3NHCH2CH2NH2
Formula Weight: 116.21
Flash Point: 59°(138°F)
Refractive Index: 1.4430
Boiling Point: 170-171°
Beilstein Reference: 1732700
UN No.: 2734
MDL No.: MFCD00025602
Formula:
C₆H₁₆N₂ Boiling Pt: 170…171 °C Density: 0,836 Flash Pt: 59 °C (138 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00025602 CAS Number: 19522-69-1 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...