2-Cyclopropylethanol 96%

Supplier: Thermo Fisher Scientific
Warning

L11080.03 L11080.06 L11080.MD
L11080.03EA 95.4 GBP
L11080.03 L11080.06 L11080.MD
2-Cyclopropylethanol 96%
2-Cyclopropylethanol

Purity: 96% CAS No.: 2566-44-1 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: 47°(116°F) Refractive Index: 1.4355 Boiling Point: 137-138° Beilstein Reference: 2036028 UN No.: 1987 MDL No.: MFCD00040762

Formula: C₅H₁₀O
Boiling Pt: 137…138 °C
Flash Pt: 47 °C (116 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040762
CAS Number: 2566-44-1
UN: 1987
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR