1-(2-Hydroxyphenyl)-2-thiourea ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: N-(2-Hydroxyphenyl)thiourea, 2-Hydroxyphenylthiourea

L10519.03 L10519.06
L10519.03EA 37 GBP
L10519.03 L10519.06
1-(2-Hydroxyphenyl)-2-thiourea ≥97%
1-(2-Hydroxyphenyl)-2-thiourea

Purity: 97% CAS No.: 1520-26-9 Molecular Formula: C7H8N2OS Formula Weight: 168.22 Odour: Odourless Melting Point: ca 158° dec. Beilstein Reference: 1102479 MDL No.: MFCD00041159

Formula: (C₆H₄(OH))NC(S)NH₂
MW: 168,22 g/mol
Boiling Pt: 323 °C (1013 hPa)
Melting Pt: ∼ 158 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00041159
CAS Number: 1520-26-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR