1,3-Dibutylurea ≥98%

Supplier: Thermo Scientific

Synonyms: N,N'-Dibutylurea, N,N'-Di-n-butylurea

L06165.06 L06165.14
L06165.06EA 63.4 GBP
L06165.06 L06165.14
1,3-Dibutylurea ≥98%
1,3-Dibutylurea

Purity: 98% CAS No.: 1792-17-2 Molecular Formula: CH3(CH2)3NHCONH(CH2)3CH3 Formula Weight: 172.27 Melting Point: 69-75° Boiling Point: 115°/0.03mm MDL No.: MFCD00015229

Formula: H₃CCH₂CH₂CH₂NHC(O)NHCH₂CH₂CH₂CH₃
MW: 172,27 g/mol
Boiling Pt: 115 °C (0,03 mmHg)
Melting Pt: 69…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00015229
CAS Number: 1792-17-2
EINECS: 217-258-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR