1-Propyl-4-iodobenzene ≥97%
Supplier: Thermo Fisher Scientific
Synonyms:
1-(4-Iodophenyl)propane, 4-Propyliodbenzol, 4-Propyliodobenzene, 1-Iodo-4-propylbenzene
L05767.06
L05767.14
L05767.06EA
70.3
GBP
L05767.06
L05767.14
1-Propyl-4-iodobenzene ≥97%
1-Propyl-4-iodobenzene
Purity: 97%
CAS No.: 126261-84-5
Molecular Formula: C9H11I
Formula Weight: 246.09
Refractive Index: 1.5790
Boiling Point: 240-242°
MDL No.: MFCD00051612
Formula:
C₉H₁₁I Boiling Pt: 240…242 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051612 CAS Number: 126261-84-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...