N-Methylformamide ≥99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Formmethylamide, Formic acid methylamide
L03908.18
L03908.30
L03908.0B
L03908.18EA
20.1
GBP
L03908.18
L03908.30
L03908.0B
N-Methylformamide ≥99%
N-Methylformamide
Purity: 99%
CAS No.: 123-39-7
Molecular Formula: C2H5NO
Formula Weight: 59.07
Flash Point: 98°(208°F)
Refractive Index: 1.4320
Melting Point: -4°
Boiling Point: 181-185°
Beilstein Reference: 1098352
Merck Reference: 14,6076
MDL No.: MFCD00003280
Formula:
C₂H₅NO MW: 59,07 g/mol Boiling Pt: 181…185 °C Melting Pt: –4 °C Density: 1,018 Flash Pt: 98 °C (208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003280 CAS Number: 123-39-7 EINECS: 204-624-6 Merck Index: 14,6076 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...