1-Naphthylmethylamine ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
1-Naphthalenemethylamine, Naphthalen-1-ylmethanamine, Naphthalene-1-methylamine, 1-(Aminomethyl)naphthalene
L03541.03
L03541.06
L03541.14
L03541.03EA
23.1
GBP
L03541.03
L03541.06
L03541.14
1-Naphthylmethylamine ≥98%
1-Naphthylmethylamine
Purity: 98+%
CAS No.: 118-31-0
Molecular Formula: C11H11N
Formula Weight: 157.22
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₁₁H₁₁N MW: 157,22 g/mol Boiling Pt: 289…292 °C Melting Pt: 267…269 °C Density: 1,075 Flash Pt: >110 °C (230 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00004048 CAS Number: 118-31-0 EINECS: 204-244-0 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...