5-Bromo-2,4-dichloropyrimidine ≥98%

Supplier: Thermo Scientific
Danger

B25365.03 B25365.06
B25365.03EA 25.8 GBP
B25365.03 B25365.06
5-Bromo-2,4-dichloropyrimidine ≥98%
5-Bromo-2,4-dichloropyrimidine

Purity: 98% CAS No.: 36082-50-5 Molecular Formula: C4HBrCl2N2 Formula Weight: 227.88 Flash Point: 110°(230°F) Refractive Index: 1.6030 Melting Point: 29-30° Boiling Point: 128°/15mm Beilstein Reference: 124441 UN No.: 2923 MDL No.: MFCD00127818

Formula: C₄HBrCl₂N₂
MW: 227,87 g/mol
Boiling Pt: 128 °C (15 mmHg)
Melting Pt: 29…30 °C
Density: 1,781
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00127818
CAS Number: 36082-50-5
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR