Butyl chloroformate ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Chloroformic acid butyl ester, n-Butyl chloroformate
B22215.0B
B22215.22
B22215.30
B22215.0BEA
124
GBP
B22215.0B
B22215.22
B22215.30
Butyl chloroformate ≥98%
Butyl chloroformate
Purity: 98%
CAS No.: 592-34-7
Molecular Formula: ClCO2CH2CH2CH2CH3
Formula Weight: 136.58
Storage Temperature: 2°C to 8°C
Flash Point: 39°(102°F)
Refractive Index: 1.4130
Odour: Acrid
Melting Point:
Formula:
C₅H₉ClO₂ MW: 136,58 g/mol Boiling Pt: 141…143 °C Melting Pt: < –70 °C Density: 1,060 Flash Pt: 39 °C (102 °F) |
MDL Number:
MFCD00000650 CAS Number: 592-34-7 EINECS: 209-750-5 UN: 2743 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...