Isophthalaldehyde ≥98%

Supplier: Thermo Scientific

Synonyms: m-Phthalaldehyde, Isophthaldicarboxaldehyde, Isophthaldialdehyde

A15788.14 A15788.06 A15788.22
A15788.14EA 117 GBP
A15788.14 A15788.06 A15788.22
Isophthalaldehyde ≥98%
Isophthalaldehyde

Purity: 98% CAS No.: 626-19-7 Molecular Formula: C8H6O2 Formula Weight: 134.13 Melting Point: 85-91° Boiling Point: 136°/17mm MDL No.: MFCD00003372

Formula: C₈H₆O₂
MW: 134,13 g/mol
Boiling Pt: 136 °C (17 mmHg)
Melting Pt: 85…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00003372
CAS Number: 626-19-7
EINECS: 210-935-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR