4-Pyridylmethanol ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Pyridylmethanol, 4-(Hydroxymethyl)pyridine, 4-Pyridinemethanol, Pyridine-4-methanol, 4-Pyridylcarbinol
A14698.14
A14698.22
A14698.14EA
46.3
GBP
A14698.14
A14698.22
4-Pyridylmethanol ≥99%
4-Pyridylmethanol
Purity: 99%
CAS No.: 586-95-8
Molecular Formula: C6H7NO
Formula Weight: 109.13
Storage Temperature: 2°C to 8°C
Flash Point: 148°(298°F)
Melting Point: 53-55°
Boiling Point: 141°/12mm
Beilstein Reference: 107850
MDL No.: MFCD00006442
Formula:
C₆H₇NO MW: 109,13 g/mol Boiling Pt: 141 °C (12 mmHg) Melting Pt: 53…55 °C Density: 1,15 g/cm³ (20 °C) Flash Pt: 148 °C (298 °F) |
MDL Number:
MFCD00006442 CAS Number: 586-95-8 EINECS: 209-590-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...