N-(4-Bromobutyl)phthalimide ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: 2-(4-Bromobutyl)isoindoline-1,3-dione

A14517.06 A14517.14 A14517.22
A14517.06EA 48.9 GBP
A14517.06 A14517.14 A14517.22
N-(4-Bromobutyl)phthalimide ≥98%
N-(4-Bromobutyl)phthalimide

Purity: 98% CAS No.: 5394-18-3 Molecular Formula: C12H12BrNO2 Formula Weight: 282.14 Odour: Odourless Melting Point: 77-81° Boiling Point: 165-170°/1mm Beilstein Reference: 164119 MDL No.: MFCD00005905

Formula: C₁₂H₁₂BrNO₂
Boiling Pt: 165…170 °C (1 mmHg)
Melting Pt: 77…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00005905
CAS Number: 5394-18-3
EINECS: 226-401-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR