6-Ethyl-o-toluidine ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Ethyl-6-methylaniline, 2-Amino-3-ethyltoluene
A12796.0E
A12796.36
A12796.0EEA
226
GBP
A12796.0E
A12796.36
6-Ethyl-o-toluidine ≥98%
6-Ethyl-o-toluidine
Purity: 98%
CAS No.: 24549-06-2
Molecular Formula: C9H13N
Formula Weight: 135.21
Flash Point: 89°(192°F)
Refractive Index: 1.5520
Melting Point: -25°
Boiling Point: 226-228°
Beilstein Reference: 2079468
MDL No.: MFCD00007752
Formula:
C₉H₁₃N MW: 135,21 g/mol Boiling Pt: 226…228 °C Melting Pt: –25 °C Density: 0,969 Flash Pt: 89 °C (192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007752 CAS Number: 24549-06-2 EINECS: 246-309-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...