4-Butoxyphenol ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Hydroquinone monobutyl ether

A12076.22 A12076.06 A12076.14
A12076.22EA 214 GBP
A12076.22 A12076.06 A12076.14
4-Butoxyphenol ≥98%
4-Butoxyphenol

Purity: 98% CAS No.: 122-94-1 Molecular Formula: C10H14O2 Formula Weight: 166.22 Melting Point: 63-66° MDL No.: MFCD00002336

Formula: C₁₀H₁₄O₂
MW: 166,22 g/mol
Boiling Pt: 125 °C (4 mmHg)
Melting Pt: 63…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00002336
CAS Number: 122-94-1
EINECS: 204-583-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR