Valeronitrile ≥99%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Pentanenitrile

A10306.AC A10306.AE A10306.AP
A10306.ACEA 24 GBP
A10306.AC A10306.AE A10306.AP
Valeronitrile ≥99%
Valeronitrile

Purity: 99% CAS No.: 110-59-8 Molecular Formula: CH3(CH2)3CN Formula Weight: 83.13 Flash Point: 38°(100°F) Refractive Index: 1.3970 Odour: Odourless Melting Point: -96° Boiling Point: 139-141° Beilstein Reference: 1736706 Merck Reference: 14,9905 UN No.: 1992 MDL No.: MFCD00001974

Formula: C₅H₉N
MW: 83.13 g/mol
Boiling Pt: 139…141 °C
Melting Pt: –96 °C
Density: 0.797
Flash Pt: 38 °C (100 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001974
CAS Number: 110-59-8
EINECS: 203-781-8
ADR: 3 III
Merck Index: 12,10043

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR