N-Methylformamide 99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Formmethylamide, Formic acid methylamide
414330050
414331000
414330010
ACRO414330050EA
32.6
GBP
ACRO414330050
ACRO414331000
ACRO414330010
N-Methylformamide 99%
N-Methylformamide
Formula:
C₂H₅NO MW: 59,07 g/mol Boiling Pt: 199…201 °C (760 mmHg) Melting Pt: –3,2 °C Density: 1,003 g/cm³ Flash Pt: 111 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003280 CAS Number: 123-39-7 EINECS: 204-624-6 Merck Index: 13,00061 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4300 to 1.4340 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...