4-Phenylbenzyl bromide 96%

Supplier: Thermo Scientific
Danger

Synonyms: 4-(Bromomethyl)biphenyl, 4-(Bromomethyl)-1,1'-biphenyl, p-(Bromomethyl)biphenyl

368950050 368950250
ACRO368950050EA 162 GBP
ACRO368950050 ACRO368950250
4-Phenylbenzyl bromide 96%
4-Phenylbenzyl bromide
Formula: C₁₃H₁₁Br
MW: 247,13 g/mol
Melting Pt: 83…86 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00017869
CAS Number: 2567-29-5
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Melting point 83 to 86 °C
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR