Iodotrimethylsilane 95-97% stabilised

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Trimethyliodosilane, Trimethylsilyl iodide

187490050 187490250 187491000 187492500
187490050.EA 77.3 GBP
187490050. 187490250. 187491000. 187492500.
Iodotrimethylsilane 95-97% stabilised
Iodotrimethylsilane
Formula: C₃H₉ISi
MW: 200.09 g/mol
Boiling Pt: 106 °C (760 mmHg)
Density: 1.4 g/cm³
Flash Pt: –31 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001028
CAS Number: 16029-98-4
EINECS: 240-171-0
UN: 2924
ADR: 3 II

Order Now

Specification Test Results

Appearance Clear colorless to red Liquid
Infrared spectrum Conforms
Refractive index 1.4700 to 1.4800 (20°C, 589 nm)
Titration Argentometric ≥95 % (Ex I)
Stabilizer copper


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR