3-Pyridylmethanol 98%

Supplier: Thermo Scientific
Warning

Synonyms: 3-(Hydroxymethyl)pyridine, 3-Pyridinemethanol, 3-Pyridylcarbinol, Pyridine-3-methanol, 3-Pyridylmethanol

132000050 132001000
ACRO132000050EA 41.9 GBP
ACRO132000050 ACRO132001000
3-Pyridylmethanol 98%
3-Pyridylmethanol
Formula: C₆H₇NO
MW: 109,13 g/mol
Boiling Pt: 259 °C (760 mmHg)
Melting Pt: –7 °C
Density: 1,124 g/cm³
Flash Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00006407
CAS Number: 100-55-0
EINECS: 202-864-6
Merck Index: 12,06614

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5425 to 1.5455 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR