Acetic acid-[D1]

Supplier: Merck
Danger

Synonyms: Ethanoic acid-D1

8.15035.0050
8.15035.0050EA 0 GBP
8.15035.0050
Acetic acid-[D1]
Acetic acid-[D1]
Formula: C₂H₃DO₂
MW: 61.04 g/mol
Boiling Pt: 116
Melting Pt: 17
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00051050
CAS Number: 758-12-3
EINECS: 212-059-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR