Steel Ruler, Series 182
Supplier: CAULFIELD INDUSTRIAL LTD
182-305
CAUL182-305EA 40.1 GBP
CAUL182-305
Steel Ruler, Series 182
Rulers
This is semi-flexible ruler, engraved on the front side only.
  • Clear graduations on satin-chrome finish
  • Stainless tempered
Graduations: ¹/₁₆", ¹/₃₂", ¹/₆₄", 1 mm, 0,5 mm.