3-Chloro-2-fluorobenzylamine, Technical Grade

Supplier: Apollo Scientific

Synonyms: (3-Chloro-2-fluorophenyl)methanamine

PC57807-5G PC57807-10G PC57807-25G
APOSPC57807-5GEA 34.1 GBP
APOSPC57807-5G APOSPC57807-10G APOSPC57807-25G
3-Chloro-2-fluorobenzylamine, Technical Grade
3-Chloro-2-fluorobenzylamine
Formula: C₇H₇ClFN
MW: 159.59 g/mol
Boiling Pt: 211
Melting Pt: 60
MDL Number: MFCD01631391
CAS Number: 72235-55-3
UN: 2735

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR