Antimicrobial plus foam handwash, GOJO®
Supplier: GOJO
GOJO®
115-3177EA 76.3 GBP
115-3177
Antimicrobial plus foam handwash, GOJO®
The quat-free GOJO® Antimicrobial Plus Foam Handwash is gentle on skin and formulated to be highly effective.
  • Suitable for sensitive skin and formulated for high frequency use
  • Enriched with a moisturising plant-based agent
  • Broad spectrum efficacy (bacteria, yeasts, and virus)
  • Unfragranced and colourless
  • Use with the GOJO® ADX-12 dispenser
Suitable for use in food handling areas such as restaurants, front and back offices, kitchens, food processing, and facility management. This formulation targets food specific pathogens such as Salmonella, Listeria, and E.coli. Food taint test compliant to BS EN ISO 4120:2007. Passes EN 1499 in 30 seconds.