2-Naphthyl trifluoromethanesulphonate 97%

Supplier: Thermo Scientific
Danger

Synonyms: 2-Naphthyl Triflate, 2-Naphthyl Trifluoromethanesulphonate, Trifluoromethanesulphonic acid 2-Naphthyl Ester

H55417.06 H55417.03 H55417.MD
H55417.06EA 187 GBP
H55417.06 H55417.03 H55417.MD
2-Naphthyl trifluoromethanesulphonate 97%
2-Naphthyl trifluoromethanesulphonate
Formula: C₁₁H₇F₃O₃S
MW: 276,23 g/mol
Melting Pt: 30…32 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00192341
CAS Number: 3857-83-8
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR