Imidacloprid

Supplier: Thermo Fisher Scientific
Warning

466750010 466752500
ACRO466750010EA 364 GBP
ACRO466750010 ACRO466752500
Imidacloprid
Imidacloprid
Formula: C₉H₁₀ClN₅O₂
MW: 255.66 g/mol
Melting Pt: 134°C…146°C
Storage Temperature: Ambient
MDL Number: MFCD00468059
CAS Number: 138261-41-3
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to light yellow to light beige crystalline powder
Infrared spectrum Conforms
Melting point 141°C to 146°C
Loss on drying ≤ 1% (1g, 105°C)
HPLC ≥ 94%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR