Rotigotine
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
(S)-6-[Propyl[2-(thiophen-2-yl)ethyl]amino]-5,6,7,8-tetrahydronaphthalen-1-ol
466541000
466545000
ACRO466541000EA
326
GBP
ACRO466541000
ACRO466545000
Rotigotine
Rotigotine
Formula:
C₁₉H₂₅NOS MW: 315,48 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00870193 CAS Number: 99755-59-6 |
Specification Test Results
Appearance | White to almost white powder |
Infrared spectrum | Conforms |
HPLC | ≥ 98% |
Titration Total base | 98 to 102% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...