Melanin (synthetic)

Supplier: Thermo Fisher Scientific

466430010 466430050 466432500
ACRO466430010EA 244 GBP
ACRO466430010 ACRO466430050 ACRO466432500
Melanin (synthetic)
Melanin
Formula: C₁₈H₁₀N₂O₄
Storage Temperature: Freezer
MDL Number: MFCD00131581
CAS Number: 8049-97-6
EINECS: 232-473-6

Order Now

Specification Test Results

Appearance Brown to black powder
Infrared spectrum Conforms
Carbon (C) 44 to 55%
Nitrogen (N) 6 to 7.5%
Solubility (20mg/ml NH4OH 1N) Brown to very dark black
Residue after ignition ≤ 2% Ash


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR