Aprepitant
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
5-(((2R,3S)-2-((R)-1-(3,5-Bis(trifluoromethyl)phenyl)ethoxy)-3-(4-fluorophenyl)morpholino)methyl)-1H-1,2,4-triazol-3(2H)-one
465310010
465312500
ACRO465310010EA
742
GBP
ACRO465310010
ACRO465312500
Aprepitant
Aprepitant
Formula:
C₂₃H₂₁F₇N₄O₃ MW: 534.43 g/mol Melting Pt: 255°C Storage Temperature: Ambient |
MDL Number:
MFCD12912491 CAS Number: 170729-80-3 UN: 3077 ADR: 9 III |
Specification Test Results
Appearance | White to almost white powder |
Infrared spectrum | Conforms |
Melting point | 250°C to 255°C |
Water | ≤ 0.5% |
HPLC | ≥ 97.5% |
Specific optical rotation | +66° to +71° (20°C, 589nm) (1% in M%eOH) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...