Sodium dithionite ≥85-87%, GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Sodium hydrosulphite

27884.260EA 51.7 GBP
27884.260 27884.290 27884.320
Sodium dithionite ≥85-87%, GPR RECTAPUR®
Sodium dithionite
Formula: Na₂S₂O₄
MW: 174,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011640
CAS Number: 7775-14-6
EINECS: 231-890-0
UN: 1384
ADR: 4.2,II
REACH: 01-2119520510-57
Merck Index: 13,08700

Order Now

Specification Test Results

Assay (iodometric) (at time of batch analysis) 85 - 87%
Iron (Fe) max. 0,002%
Appearance white crystalline powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR