Masterflex® Bag Filters for Polypropylene Bag Filter Housings, Avantor®
Supplier: Avantor Fluid Handling
01516-66
MFLX01516-66EA 27.7 GBP
MFLX01516-66
Masterflex® Bag Filters for Polypropylene Bag Filter Housings, Avantor®
Filters Bag Filter Systems
Conveniently collect all filtered particles within the bag.
  • Choose from a wide range of bag materials suitable for your application
Choose from standard felt bags, high-efficiency bags, or nylon mesh bags. Each bag measures 4" diameter. The 10"L bags have a surface area of 0.75 ft2; 18-½"L bags have a surface area of 1.75 ft2.