Water purification systems, Elix®
Supplier: Merck
ZRX0003BE ZRX0003FR
171-0396EA 0 GBP
171-0396 171-0379
Water purification systems, Elix®
Water Purification Systems Pure and Ultra Pure Water Systems
Elix® systems provide pure water of consistent and reliable quality. They allow the user to make real savings. Combining Millipore’s patented Elix electrodeionisation technology with the most advanced purification technologies, the Elix® system produces top quality pure water from tap water and satisfies all your laboratory needs. Produces Type II purified water from tap water.
  • Operating costs are reduced, because the system uses electricity to continuously regenerate the resins and remove ions from the tap water. There is no need to replace or regenerate the resin cartridges
  • Evolutionary flow equipment: the Elix® 3 (water production 3 L/h) adapts to the user's needs and can be transformed into models 5, 10 or 15 (up to 15 L/h)
  • Advanced reverse osmosis guarantees a constant flow of product water
  • The waste recirculation loop helps save tap water
  • Complete process monitoring system systematically checks water temperature, pressure, conductivity and ion rejection using sensors calibrated to the different stages of purification
The pure water produced by the Elix® system can be used in laboratory applications, such as the preparation of reagents and buffers, the preparation of microbiological cultures, histology applications and as purified water for pharmaceutical applications (conforms to American and European pharmacopeias). It is also ideal for feeding laboratory equipment (washing machines, clinical analysers, stability test cabinets, humidifiers, autoclaves and climatic cabinets). Elix water is recommended for feeding all ultrapure water production systems.
Full Quality Assurance: conformity certificates and quality, calibration and qualification documents are available on request.
Full Quality Assurance: conformity certificates and quality, calibration and qualification documents are available on request.
With Ethernet connection (RJ45) for data transfer. Elix units are supplied without purification pack.
Delivery information: With Ethernet connection (RJ45) for data transfer. Elix units are supplied without purification pack.