Mammalian TAP system, interPlay
Supplier: AGILENT
240102 240103 240104 240106
AGLS240102EA 663 GBP
AGLS240102 AGLS240103 AGLS240104 AGLS240106
Mammalian TAP system, interPlay
Vectors, Plasmids and Libraries
The InterPlay Mammalian TAP system allows you to recover interacting proteins from mammalian cells.
  • Protease digestion unnecessary to recover intact interacting protein partners since SBP and CBP elute easily from their affinity resins
  • Exceptionally pure protein for downstream analysis
This method is based on expression of a protein of interest fused to two affinity tags: a streptavidin binding peptide (SBP) and a calmodulin binding peptide (CBP). Tandem affinity purification yields your tagged protein and interacting proteins using gentle washing and small molecule elution conditions.
Caution: For research use only. Not for use in diagnostic procedures.