pBC phagemid vectors
Supplier: AGILENT
212215 212216 212217 212218
AGLS212215EA 520 GBP
AGLS212215 AGLS212216 AGLS212217 AGLS212218
pBC phagemid vectors
Vectors, Plasmids and Libraries
The pBC vectors were derived from the pBluescript II phagemid. The ampicillin-resistance gene has been replaced with the chloramphenicol resistance gene.
  • Convenient selection when subcloning from ampicillin-resistant cloning vectors
  • Large and versatile polylinker in two orientations for cloning flexibility
  • Available with f1 origin in either (+) or (–) orientation
pBC phagemids (plasmids with a phage origin) are cloning vectors designed to simplify commonly used cloning and sequencing procedures, including the construction of nested deletions for DNA sequencing, generation of RNA transcripts in vitro and site-specific mutagenesis and gene mapping.
Caution: For research use only. Not for use in diagnostic procedures.