Capsimycin
Supplier: Apollo Scientific
BIBR1142-1MG
BIBR1142-5MG
APOSBIBR1142-1MGEA
276
GBP
APOSBIBR1142-1MG
APOSBIBR1142-5MG
Capsimycin
Capsimycin
From microbial source
Storage Temperature:
Ambient |
CAS Number:
70694-08-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...