2-Chloro-N-(2-fluorophenyl)acetamide

Supplier: Apollo Scientific
Warning

Synonyms: N-(Chloroacetyl)-2-fluoroaniline

PC9999-1G PC9999-5G
APOSPC9999-1GEA 19.7 GBP
APOSPC9999-1G APOSPC9999-5G
2-Chloro-N-(2-fluorophenyl)acetamide
2-Chloro-N-(2-fluorophenyl)acetamide
Formula: C₈H₇ClFNO
MW: 187,6 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00730970
CAS Number: 347-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR