2,2-Difluoro-2-(fluorosulphonyl)acetic acid 98%
Supplier: Apollo Scientific
|
Danger
|
PC9975-100G
PC9975-25G
APOSPC9975-100GEA
124
GBP
APOSPC9975-100G
APOSPC9975-25G
2,2-Difluoro-2-(fluorosulphonyl)acetic acid 98%
2,2-Difluoro-2-(fluorosulphonyl)acetic acid
Methyl and trimethylsilyl esters also available.
Formula:
C₂HF₃O₄S Boiling Pt: 153 °C Density: 1,723 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01320807 CAS Number: 1717-59-5 UN: 2927 ADR: 6.1+8,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...