4-Fluoropiperidine hydrochloride 95%

Supplier: Apollo Scientific
Danger

PC9784-1G PC9784-25G PC9784-5G
APOSPC9784-1GEA 22.3 GBP
APOSPC9784-1G APOSPC9784-25G APOSPC9784-5G
4-Fluoropiperidine hydrochloride 95%
4-Fluoropiperidine hydrochloride
Formula: C₅H₁₁ClFN
Melting Pt: 161…167 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03452786
CAS Number: 57395-89-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR