2-Amino-5-fluorophenol 97%

Supplier: Apollo Scientific
Warning

PC9625-25G PC9625-5G PC9625-100G
APOSPC9625-25GEA 53.7 GBP
APOSPC9625-25G APOSPC9625-5G APOSPC9625-100G
2-Amino-5-fluorophenol 97%
2-Amino-5-fluorophenol
Formula: C₆H₆FNO
Melting Pt: 135…140 °C
Storage Temperature: Ambient
MDL Number: MFCD00671759
CAS Number: 53981-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR