3-Isocyanato-1-(trifluoroacetyl)piperidine 95+%

Supplier: Apollo Scientific
Danger

PC9567-1G PC9567-5G PC9567-10G
APOSPC9567-1GEA 315 GBP
APOSPC9567-1G APOSPC9567-5G APOSPC9567-10G
3-Isocyanato-1-(trifluoroacetyl)piperidine 95+%
3-Isocyanato-1-(trifluoroacetyl)piperidine

  • May form small amount of sediment.

Formula: C₈H₉F₃N₂O₂
Boiling Pt: 129…132 °C/5,25mmHg
Storage Temperature: Ambient
MDL Number: MFCD03424489
CAS Number: 808764-41-2
UN: 2206
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR