m,α,α,α-Tetrafluoroanisole 99%

Supplier: Apollo Scientific
Warning

PC9545-25G PC9545-5G
APOSPC9545-25GEA 61.7 GBP
APOSPC9545-25G APOSPC9545-5G
m,α,α,α-Tetrafluoroanisole 99%
m,α,α,α-Tetrafluoroanisole
Formula: C₇H₄F₄O
MW: 180,1 g/mol
Boiling Pt: 100…105 °C
Density: 1,33 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236323
CAS Number: 1077-01-6
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR