6-Fluoroindole 98%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Fluoro-1H-indole

PC9422-100G PC9422-25G
APOSPC9422-100GEA 187 GBP
APOSPC9422-100G APOSPC9422-25G
6-Fluoroindole 98%
6-Fluoroindole
Formula: C₈H₆FN
MW: 135,14 g/mol
Melting Pt: 72…77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00056933
CAS Number: 399-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR