Methyl-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylate 97%

Supplier: Apollo Scientific

PC9403-1G PC9403-5G
APOSPC9403-1GEA 159 GBP
APOSPC9403-1G APOSPC9403-5G
Methyl-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylate 97%
Methyl-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylate
Formula: C₁₃H₁₄ClF₃N₂O₂
MW: 322,72 g/mol
Melting Pt: 52…54 °C
Storage Temperature: Ambient
MDL Number: MFCD00172275
CAS Number: 321430-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR