4-Amino-6-(trifluoromethyl)quinoline

Supplier: Apollo Scientific
Warning

Synonyms: 6-(Trifluoromethyl)quinolin-4-amine

PC9212-500MG PC9212-250MG
APOSPC9212-500MGEA 0 GBP
APOSPC9212-500MG APOSPC9212-250MG
4-Amino-6-(trifluoromethyl)quinoline
4-Amino-6-(trifluoromethyl)quinoline
Formula: C₁₀H₇F₃N₂
MW: 212,17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00278781
CAS Number: 247113-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR